Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073206.1 | 17 | 1652 | |||
XM_006723646.2 | XP_006723709.1 | 16 | 1533 | |||
XM_006723647.2 | XP_006723710.1 | 16 | 1533 | |||
XM_006723648.2 | XP_006723711.1 | 15 | 1476 | |||
XM_006723650.2 | XP_006723713.1 | 16 | 1533 | |||
XM_006723651.2 | XP_006723714.1 | 16 | 1533 | |||
XM_006723654.3 | XP_006723717.1 | 7 | 548 | |||
XM_011529374.1 | XP_011527676.1 | 15 | 1473 | |||
XM_011529375.1 | XP_011527677.1 | 16 | 1422 | |||
XM_011529376.1 | XP_011527678.1 | 15 | 1365 | |||
XM_011529377.1 | XP_011527679.1 | 17 | 1742 | |||
XM_011529378.1 | XP_011527680.1 | 14 | 1251 | |||
XM_011529379.1 | XP_011527681.1 | 14 | 1284 | |||
XM_017028084.1 | XP_016883573.1 | 5 | 470 |