Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316964.1 | NP_001303893.1 | 4 | 729 | ||
NM_001316965.1 | NP_001303894.1 | 4 | 729 | |||
NM_025232.3 | NP_079508.2 | 4 | 729 | |||
GenBank mRNA | AF218014.1 | - | 4 | 507 | ||
AK025899.1 | - | 4 | 659 | |||
AK025930.1 | - | 4 | 362 | |||
AK172846.1 | - | 4 | 664 | |||
AK302352.1 | - | 4 | 546 | |||
AY562242.1 | - | 4 | 260 | |||
BC013048.1 | - | 4 | 713 | 3863838 | ||
BC050622.1 | - | 4 | 631 | 6504298 | ||
BG695963.1 | - | 4 | 318 | |||
CR457353.1 | - | 4 | 260 |