Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030572.3 | NP_085049.1 | 4, 5 | 425 | ||
NR_135187.1 | 1, 2 | 356 | ||||
GenBank mRNA | AK027273.1 | - | 4, 5 | 263 | ||
AK075342.1 | - | 4, 5 | 379 | |||
AK311147.1 | - | 1, 2 | 126 | |||
BC004336.1 | - | 4, 5 | 360 | 3631700 | ||
DA260291.1 | - | 1, 2 | 356 | |||
DA380664.1 | - | 4, 5 | 424 |