Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145165.1 | NP_001138637.1 | 4 | 977 | ||
NM_031304.4 | NP_112594.1 | 4 | 688 | |||
GenBank mRNA | BC002817.2 | - | 4 | 688 | 3635058 | |
BC009863.2 | - | 3 | 577 | 3938535 |