Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
Gene Name:
BCL2 like 12
Gene Aliases:
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
siRNA ID
HSS149674
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
1
757
NM_001282516.1
NP_001269445.1
1
757
NM_001282517.1
NP_001269446.1
1
757
NM_001282519.1
NP_001269448.1
1
757
NM_001282520.1
NP_001269449.1
1
757
NM_001282521.1
NP_001269450.1
1
757
NM_138639.1
NP_619580.1
1
757
NR_104200.1
1
757
NR_104201.1
1
757
NR_104202.1
1
757
NR_104203.1
1
757
NR_104204.1
1
757
NR_104205.1
1
757
XM_017027345.1
XP_016882834.1
1
827
XM_017027346.1
XP_016882835.1
1
827
GenBank mRNA
BC104004.1
-
1
333
40024338
BC104005.1
-
1
333
40024342
BC104006.1
-
1
333
40024345
FJ868798.1
-
1
75
FJ868799.1
-
1
75
FJ868800.1
-
1
75
FJ868801.1
-
1
75
FJ868802.1
-
1
75
FJ868803.1
-
1
75
FJ911906.1
-
1
75
FJ911907.1
-
1
75
FJ911908.1
-
1
75
HM347053.1
-
1
75
KR820244.1
-
1
75
KR820245.1
-
1
75
KR820246.1
-
1
75
KR820247.1
-
1
75
KR820248.1
-
1
75
KR820249.1
-
1
75
KR820250.1
-
1
75
KR820251.1
-
1
75
KR820252.1
-
1
75
KR820253.1
-
1
75
KR820254.1
-
1
75
KR820255.1
-
1
75
KR820256.1
-
1
75
KR820257.1
-
1
75
KR820258.1
-
1
75
KR820259.1
-
1
75
KR820260.1
-
1
75
KR820261.1
-
1
75
KR820262.1
-
1
75
KR820263.1
-
1
75
KR820264.1
-
1
75
KR820265.1
-
1
75
KR820266.1
-
1
75
KR820267.1
-
1
75
KR820268.1
-
1
75
KR820269.1
-
1
75
KR820270.1
-
1
75
KR820271.1
-
1
75
KR820272.1
-
1
75
KR820273.1
-
1
75
KR820274.1
-
1
75
KR820275.1
-
1
75
KR820276.1
-
1
75
KR820277.1
-
1
75
KR820278.1
-
1
75
KR820279.1
-
1
75
KR820280.1
-
1
75
KR820281.1
-
1
75
KR820282.1
-
1
75
KR820283.1
-
1
75
KR820284.1
-
1
75
KR820285.1
-
1
75
KR820286.1
-
1
75
KR820287.1
-
1
75
KR820288.1
-
1
75
KR820289.1
-
1
75
KR820290.1
-
1
75
KR820291.1
-
1
75
KR820292.1
-
1
75
KR820293.1
-
1
75
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0