Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032199.2 | NP_115575.1 | 4, 5 | 1188 | ||
GenBank mRNA | AK296921.1 | - | 4, 5 | 754 | ||
BC107800.1 | - | 4, 5 | 747 |