Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032214.3 | NP_115590.1 | 5 | 727 | ||
NM_175077.2 | NP_778252.1 | 5 | 727 | |||
XM_017028098.1 | XP_016883587.1 | 5 | 718 | |||
GenBank mRNA | AF290985.1 | - | 4 | 304 | ||
AF290986.1 | - | 4 | 304 | |||
AF326353.1 | - | 4 | 304 | |||
AK025645.1 | - | 5 | 364 | |||
AK291513.1 | - | 5 | 707 | |||
BC042041.1 | - | 5 | 666 | 4429896 | ||
BQ052468.1 | - | 5 | 392 |