Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLYWCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308068.1 | NP_001294997.1 | 6 | 1674 | ||
NM_020912.1 | NP_065963.1 | 6 | 1671 | |||
NM_032296.2 | NP_115672.2 | 6 | 1671 | |||
XM_006720962.3 | XP_006721025.1 | 6 | 1776 | |||
XM_011522701.2 | XP_011521003.1 | 6 | 1778 | |||
XM_011522702.2 | XP_011521004.1 | 6 | 2252 | |||
XM_011522703.2 | XP_011521005.1 | 7 | 1873 | |||
XM_011522704.2 | XP_011521006.1 | 6 | 1776 | |||
XM_011522705.2 | XP_011521007.1 | 6 | 1777 | |||
XM_011522707.2 | XP_011521009.1 | 6 | 1780 | |||
XM_017023772.1 | XP_016879261.1 | 8 | 1970 | |||
GenBank mRNA | AB046772.1 | - | 6 | 1651 | ||
AK026885.1 | - | 2 | 104 | |||
AK055777.1 | - | 1 | 700 | |||
AK057022.1 | - | 2 | 1415 | |||
AK092434.1 | - | 2 | 491 | |||
AL136585.1 | - | 2 | 496 | |||
BC001973.1 | - | 3 | 599 | |||
BC018712.1 | - | 2 | 419 | 4875281 | ||
BC028572.1 | - | 6 | 1675 | 4839062 |