Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146028.1 | NP_001139500.1 | 2 | 1131 | ||
NM_032452.2 | NP_115828.2 | 3 | 1257 | |||
GenBank mRNA | AB058734.1 | - | 2 | 1131 | ||
AK091572.1 | - | 2 | 900 | |||
AK290511.1 | - | 3 | 677 | |||
BC055429.1 | - | 3 | 1242 | 5262644 |