Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032530.1 | NP_115919.1 | 2 | 2472 | ||
XM_005256827.2 | XP_005256884.1 | 2 | 2785 | |||
GenBank mRNA | AB058774.2 | - | 1 | 2457 | ||
AK131271.1 | - | 3 | 1643 | |||
BC136604.1 | - | 1 | 2450 | 9020594 |