Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1F10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032556.5 | NP_115945.4 | 2 | 519 | ||
NM_173161.2 | NP_775184.1 | 3 | 173 | |||
GenBank mRNA | AF334755.1 | - | 2 | 519 | ||
AY029413.1 | - | 3 | 173 | |||
AY358846.1 | - | 3 | 161 | |||
BC103966.1 | - | 3 | 164 | 40014540 | ||
BC103967.1 | - | 3 | 164 | 40014541 | ||
BC103968.1 | - | 3 | 164 | 40014542 |