Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 2 | 210 | ||
GenBank mRNA | AF268032.1 | - | 2 | 171 | ||
AF423421.1 | - | 2 | 144 | |||
AJ347750.1 | - | 2 | 144 | |||
AK300775.1 | - | 2 | 180 | |||
AK315105.1 | - | 2 | 180 | |||
AL831950.1 | - | 1 | 58 | |||
BC036447.2 | - | 2 | 180 | 4830913 | ||
DC348657.1 | - | 2 | 211 |