Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135111.1 | NP_001128583.1 | 4 | 777 | ||
NM_031460.3 | NP_113648.2 | 4 | 776 | |||
GenBank mRNA | AF339912.1 | - | 4 | 742 | ||
AF358910.1 | - | 4 | 636 | |||
AK310582.1 | - | 3 | 1104 | |||
AK315273.1 | - | 4 | 767 | |||
AY358853.1 | - | 4 | 727 | |||
BC025726.1 | - | 4 | 723 | 5201326 | ||
EU978944.1 | - | 4 | 683 |