Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHOT2 SIRNA Assays ›
Gene Symbol:
RHOT2
Gene Name:
ras homolog family member T2
Gene Aliases:
ARHT2, C16orf39, MIRO-2, MIRO2, RASL
Chromosome Location:
Chr.16: 668083 - 674174 on Build GRCh38
UniGene ID:
Hs.513242
Species:
Human
siRNA ID
HSS150411
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHOT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138769.2
NP_620124.1
10, 11
855
XM_005255660.2
XP_005255717.1
10, 11
809
XM_005255661.2
XP_005255718.1
9, 10
755
XM_005255662.2
XP_005255719.1
9, 10
752
XM_005255663.2
XP_005255720.1
9, 10
707
XM_006720970.2
XP_006721033.1
10, 11
792
XM_006720973.2
XP_006721036.1
9, 10
690
XM_017023825.1
XP_016879314.1
10, 11
809
XM_017023826.1
XP_016879315.1
10, 11
806
XM_017023827.1
XP_016879316.1
9, 10
755
XM_017023828.1
XP_016879317.1
9, 10
752
XM_017023829.1
XP_016879318.1
9, 10
707
XM_017023830.1
XP_016879319.1
8, 9
653
XM_017023831.1
XP_016879320.1
10, 11
792
XM_017023832.1
XP_016879321.1
7, 8
553
XM_017023833.1
XP_016879322.1
10, 11
789
XM_017023834.1
XP_016879323.1
8, 9
599
XM_017023835.1
XP_016879324.1
10, 11
804
XM_017023836.1
XP_016879325.1
9, 10
690
XM_017023837.1
XP_016879326.1
10, 11
789
XM_017023838.1
XP_016879327.1
9, 10
670
XM_017023839.1
XP_016879328.1
10, 11
804
GenBank mRNA
AL834163.1
-
9, 10
873
AY207375.1
-
10, 11
852
BC014942.1
-
10, 11
805
4859240
Back To Top
More Information
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
cellular homeostasis
mitochondrion transport along microtubule
regulation of small GTPase mediated signal transduction
mitochondrial outer membrane permeabilization
GTPase activity
calcium ion binding
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0