Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206665.2 | NP_001193594.1 | 4 | 831 | ||
NM_001320247.1 | NP_001307176.1 | 3 | 973 | |||
NM_052868.5 | NP_443100.1 | 3 | 973 | |||
XM_006711631.3 | XP_006711694.1 | 3 | 722 | |||
XM_011510159.2 | XP_011508461.1 | 3 | 1353 | |||
XM_017002835.1 | XP_016858324.1 | 4 | 837 | |||
XM_017002836.1 | XP_016858325.1 | 4 | 1028 | |||
XM_017002837.1 | XP_016858326.1 | 4 | 837 | |||
XM_017002838.1 | XP_016858327.1 | 3 | 2973 | |||
XM_017002839.1 | XP_016858328.1 | 4 | 2885 | |||
XM_017002840.1 | XP_016858329.1 | 4 | 999 | |||
GenBank mRNA | AF311906.1 | - | 3 | 802 | ||
AF407274.1 | - | 3 | 913 | |||
AK090396.1 | - | 1 | 3158 | |||
AK308115.1 | - | 4 | 833 | |||
AY044845.1 | - | 3 | 828 | |||
AY157579.1 | - | 3 | 782 | |||
BC004108.2 | - | 2 | 370 | |||
BC023649.2 | - | 1 | 38 | |||
BC053881.1 | - | 3 | 919 | 5752209 |