Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008738.2 | NP_001008738.2 | 13 | 1657 | ||
NM_133372.2 | NP_588613.2 | 14 | 1741 | |||
GenBank mRNA | AB075841.1 | - | 8 | 930 | ||
AK292473.1 | - | 14 | 1695 | |||
CR749426.1 | - | 8 | 980 | |||
DQ145719.1 | - | 14 | 1646 |