Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053046.3 | NP_444274.1 | 4 | 1478 | ||
NM_080732.3 | NP_542770.2 | 4 | 1388 | |||
GenBank mRNA | AJ310544.1 | - | 3 | 1330 | ||
AK026863.1 | - | 2 | 424 | |||
AK074408.1 | - | 4 | 1317 | |||
AK098182.1 | - | 3 | 2062 | |||
AL133009.1 | - | 1 | 3136 | |||
AL832506.1 | - | 4 | 1344 | |||
AY040565.1 | - | 3 | 1322 | |||
BC001723.1 | - | 4 | 1291 | |||
BC036051.1 | - | 4 | 1392 | 5270945 |
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 10 | 2082 | |||
GenBank mRNA | AK291385.1 | - | 10 | 2030 |