Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026547.1 | 2 | 672 | |||
GenBank mRNA | AF426269.1 | - | 2 | 672 | ||
BC101650.1 | - | 2 | 188 | |||
BC101654.1 | - | 2 | 188 |