Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080433.1 | NP_001073902.1 | 5 | 2057 | ||
XM_005264125.1 | XP_005264182.1 | 4 | 1447 | |||
GenBank mRNA | AB067499.2 | - | 5 | 2141 | ||
BC131557.1 | - | 5 | 729 |