Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052920.1 | NP_443152.1 | 11 | 2478 | ||
XM_006711929.3 | XP_006711992.1 | 10 | 3574 | |||
XM_011532501.2 | XP_011530803.1 | 8 | 1157 | |||
XM_017003264.1 | XP_016858753.1 | 10 | 2378 | |||
GenBank mRNA | AB067508.1 | - | 5 | 3781 | ||
AL834515.1 | - | 9 | 1729 | |||
BC013982.1 | - | 8 | 1467 | |||
BC015667.2 | - | 7 | 1490 |