Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146202.1 | NP_001139674.1 | 9 | 1381 | ||
NM_001146204.1 | NP_001139676.1 | 9 | 1216 | |||
NM_001146205.1 | NP_001139677.1 | 9 | 1321 | |||
NM_001146207.1 | NP_001139679.1 | 7 | 1132 | |||
NM_170604.2 | NP_733749.1 | 9 | 1423 | |||
GenBank mRNA | AF448437.1 | - | 9 | 1280 | ||
AK097292.1 | - | 2 | 351 | |||
AY048119.1 | - | 9 | 1423 | |||
AY048120.1 | - | 8 | 1538 | |||
AY048121.1 | - | 9 | 1381 | |||
BC146669.1 | - | 9 | 1263 | 40148170 | ||
BC150202.1 | - | 9 | 1263 | 40148808 | ||
BC151216.1 | - | 8 | 1432 | |||
FJ768678.1 | - | 9 | 1216 | |||
FJ768679.1 | - | 9 | 1321 | |||
FJ768681.1 | - | 9 | 1423 | |||
FJ768682.1 | - | 7 | 1132 |