Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312871.1 | NP_001299800.1 | 4 | 452 | ||
NM_001312872.1 | NP_001299801.1 | 4 | 460 | |||
NM_001312873.1 | NP_001299802.1 | 2 | 289 | |||
NM_001312874.1 | NP_001299803.1 | 3 | 420 | |||
NM_198076.5 | NP_932342.1 | 3 | 571 | |||
NR_132419.1 | 2 | 309 | ||||
NR_132420.1 | 3 | 404 | ||||
NR_132421.1 | 2 | 285 | ||||
GenBank mRNA | AF086182.1 | - | 2 | 84 | ||
AK000866.1 | - | 3 | 214 | |||
AK125259.1 | - | 3 | 563 | |||
AK130498.1 | - | 3 | 237 | |||
AK300760.1 | - | 2 | 90 | |||
AL122072.1 | - | 2 | 2340 | |||
BC018519.1 | - | 3 | 238 | 4476038 | ||
BC062419.1 | - | 3 | 239 | 6503577 | ||
BC064399.1 | - | 3 | 202 | |||
BC095486.1 | - | 3 | 208 | 30719929 | ||
BG708903.1 | - | 3 | 579 | |||
BQ189361.1 | - | 4 | 320 | |||
DA164934.1 | - | 4 | 454 | |||
DA752002.1 | - | 4 | 444 | |||
DB189037.1 | - | 3 | 220 |
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001115.1 | XP_016856604.1 | 15 | 13856 | ||
XM_017001116.1 | XP_016856605.1 | 16 | 11580 | |||
XM_017001117.1 | XP_016856606.1 | 15 | 13650 | |||
GenBank mRNA | AF448858.1 | - | 1 | 1241 | ||
AK054764.1 | - | 1 | 647 | |||
AK095297.1 | - | 2 | 3192 |