Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271907.1 | NP_001258836.1 | 2, 3 | 310 | ||
NM_001271908.1 | NP_001258837.1 | 2, 3 | 228 | |||
NM_001271909.1 | NP_001258838.1 | 1, 2 | 366 | |||
NM_153025.2 | NP_694570.1 | 2, 3 | 257 | |||
GenBank mRNA | AK056168.1 | - | 2, 3 | 199 | ||
AK303024.1 | - | 2, 3 | 276 | |||
BC148299.2 | - | 2, 3 | 199 | 40112476 | ||
BP345094.1 | - | 1, 2 | 366 | |||
DA300380.1 | - | 2, 3 | 220 | |||
DB025192.1 | - | 2, 3 | 257 | |||
DB452323.1 | - | 2, 3 | 310 | |||
DC374690.1 | - | 2, 3 | 278 |