Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284511.1 | NP_001271440.1 | 1 | 488 | ||
NM_001284512.1 | NP_001271441.1 | 1 | 488 | |||
NM_001324118.1 | NP_001311047.1 | 1 | 488 | |||
NM_152341.4 | NP_689554.2 | 1 | 488 | |||
GenBank mRNA | AK054564.1 | - | 1 | 189 | ||
AK092173.1 | - | 1 | 175 | |||
AK291373.1 | - | 1 | 190 | |||
AL832527.1 | - | 1 | 101 | |||
AY424282.1 | - | 1 | 58 | |||
BC033703.1 | - | 1 | 146 | 4154019 | ||
DA268775.1 | - | 1 | 392 | |||
DB505162.1 | - | 1 | 385 |