Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145253.2 | NP_660296.1 | 3 | 649 | ||
XM_017022937.1 | XP_016878426.1 | 3 | 548 | |||
GenBank mRNA | AF447881.1 | - | 1 | 1876 | ||
BC008681.1 | - | 3 | 604 | |||
BC012018.1 | - | 3 | 571 | 4477559 | ||
BC025327.1 | - | 3 | 592 | 3614122 | ||
BC062534.1 | - | 3 | 649 | 4620605 |