Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026865.1 | 1 | 1112 | |||
GenBank mRNA | AF063598.1 | - | 1 | 879 | ||
AF439977.1 | - | 1 | 1112 | |||
AK056371.1 | - | 1 | 1028 | |||
BC031037.1 | - | 1 | 997 | |||
BC070228.1 | - | 2 | 437 |