Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178829.4 | NP_849151.2 | 2 | 398 | ||
GenBank mRNA | AB083484.1 | - | 2 | 346 | ||
AI990561.1 | - | 1 | 373 | |||
BC014596.1 | - | 2 | 403 | 4825428 | ||
KU178863.1 | - | 2 | 282 | |||
KU178864.1 | - | 1 | 869 |