Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136273.1 | NP_001129745.1 | 3, 4 | 255 | ||
XM_005274652.3 | XP_005274709.1 | 5, 6 | 565 | |||
XM_011531115.2 | XP_011529417.1 | 4, 5 | 516 | |||
XM_017029281.1 | XP_016884770.1 | 6, 7 | 914 |