Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080621.4 | NP_542188.1 | 1 | 190 | ||
XM_005260199.4 | XP_005260256.1 | 2 | 1210 | |||
XM_017027671.1 | XP_016883160.1 | 2 | 1207 | |||
GenBank mRNA | BC067362.1 | - | 1 | 187 | 6672007 | |
BM839907.1 | - | 1 | 190 |