Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM109A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177996.1 | NP_001171467.1 | 4 | 497 | ||
NM_001177997.1 | NP_001171468.1 | 3 | 390 | |||
NM_144671.4 | NP_653272.2 | 3 | 409 | |||
XM_006719257.3 | XP_006719320.1 | 4 | 742 | |||
XM_011537976.2 | XP_011536278.1 | 2 | 216 | |||
XM_017018883.1 | XP_016874372.1 | 2 | 238 | |||
GenBank mRNA | AK093052.1 | - | 2 | 122 | ||
AK299344.1 | - | 3 | 368 | |||
BC014091.2 | - | 3 | 383 | |||
BC018996.2 | - | 3 | 383 | |||
BC034809.1 | - | 3 | 310 | 4943538 | ||
CX758147.1 | - | 4 | 497 | |||
DA707326.1 | - | 3 | 345 | |||
DB055268.1 | - | 2 | 122 |