Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM109A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177996.1 | NP_001171467.1 | 4 | 666 | ||
NM_001177997.1 | NP_001171468.1 | 3 | 559 | |||
NM_144671.4 | NP_653272.2 | 3 | 578 | |||
XM_006719257.3 | XP_006719320.1 | 4 | 911 | |||
XM_011537976.2 | XP_011536278.1 | 2 | 385 | |||
XM_017018883.1 | XP_016874372.1 | 2 | 407 | |||
GenBank mRNA | AK093052.1 | - | 2 | 291 | ||
AK299344.1 | - | 3 | 537 | |||
BC034809.1 | - | 3 | 479 | 4943538 | ||
CX758147.1 | - | 4 | 666 | |||
DA707326.1 | - | 3 | 514 | |||
DB055268.1 | - | 2 | 291 |