Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIF18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264573.1 | NP_001251503.1 | 2 | 206 | ||
NM_001265577.1 | NP_001252506.1 | 2 | 206 | |||
XM_011524385.2 | XP_011522687.1 | 2 | 507 | |||
XM_011524386.2 | XP_011522688.1 | 2 | 241 | |||
XM_011524387.2 | XP_011522689.1 | 2 | 508 | |||
XM_011524388.2 | XP_011522690.1 | 2 | 507 | |||
XM_011524389.2 | XP_011522691.1 | 2 | 507 | |||
XM_011524390.2 | XP_011522692.1 | 2 | 508 | |||
XM_011524391.2 | XP_011522693.1 | 2 | 510 | |||
GenBank mRNA | BC136590.1 | - | 1 | 132 | 9020580 | |
BC144271.1 | - | 1 | 132 | 9052791 | ||
BX405609.2 | - | 2 | 205 | |||
GU808359.1 | - | 1 | 135 |