Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541795.2 | XP_011540097.1 | 9 | 2306 | ||
XM_017001814.1 | XP_016857303.1 | 9 | 1053 | |||
XM_017001815.1 | XP_016857304.1 | 9 | 1385 | |||
XM_017001817.1 | XP_016857306.1 | 8 | 2231 | |||
XM_017001818.1 | XP_016857307.1 | 9 | 2129 | |||
XM_017001819.1 | XP_016857308.1 | 11 | 1607 | |||
XM_017001821.1 | XP_016857310.1 | 11 | 1275 | |||
XM_017001827.1 | XP_016857316.1 | 9 | 2376 | |||
XM_017001828.1 | XP_016857317.1 | 10 | 1646 |
Gene Symbol: DCDC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099434.1 | NP_001092904.1 | 1 | 245 | ||
XM_017000413.1 | XP_016855902.1 | 1 | 526 | |||
GenBank mRNA | BC041458.1 | - | 1 | 243 | ||
BC128073.1 | - | 1 | 245 | 40109068 |