Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198149.2 | NP_937792.2 | 3 | 591 | ||
NR_030775.1 | 3 | 313 | ||||
XM_017000419.1 | XP_016855908.1 | 3 | 330 | |||
GenBank mRNA | AK294106.1 | - | 3 | 347 | ||
AK316312.1 | - | 3 | 589 | |||
AY358589.1 | - | 3 | 364 | |||
BC009558.1 | - | 3 | 368 | |||
BC061908.1 | - | 2 | 210 | |||
BC104444.1 | - | 3 | 325 | 40022864 | ||
BC104445.1 | - | 3 | 325 | 40022873 | ||
BC111855.1 | - | 3 | 579 | 5762600 | ||
BM923401.1 | - | 314 |