Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178840.3 | NP_849162.1 | 2 | 501 | ||
GenBank mRNA | AI732325.1 | - | 1 | 419 | ||
AK127425.1 | - | 2 | 501 | |||
AW299952.1 | - | 1 | 430 | |||
BC017946.1 | - | 2 | 501 | 4295881 | ||
DA387602.1 | - | 2 | 501 |