Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015406.2 | 2 | 1532 | |||
GenBank mRNA | AL137476.1 | - | 1 | 672 | ||
BC067900.1 | - | 2 | 1549 |