Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284263.1 | NP_001271192.1 | 4 | 2492 | ||
NM_001284264.1 | NP_001271193.1 | 4 | 2321 | |||
NM_001284265.1 | NP_001271194.1 | 5 | 2272 | |||
NM_173510.3 | NP_775781.1 | 5 | 2546 | |||
GenBank mRNA | AK091133.1 | - | 5 | 2472 | ||
AK092636.1 | - | 1 | 365 | |||
AK127794.1 | - | 1 | 2764 | |||
BC040488.1 | - | 5 | 2502 | 5272169 | ||
BC053874.1 | - | 5 | 2505 | 5531689 |