Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138481.1 | NP_612490.1 | 3 | 539 | ||
XM_005261370.3 | XP_005261427.1 | 3 | 585 | |||
XM_011529932.2 | XP_011528234.1 | 2 | 1203 | |||
XM_011529933.2 | XP_011528235.1 | 2 | 1201 | |||
XM_011529934.2 | XP_011528236.1 | 2 | 1203 | |||
GenBank mRNA | BC040188.1 | - | 3 | 507 | ||
BC068590.1 | - | 3 | 539 |