Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120595.1 | 2 | 932 | |||
NR_120596.1 | 2 | 513 | ||||
NR_120597.1 | 2 | 170 | ||||
GenBank mRNA | AI914073.1 | - | 1 | 311 | ||
AV709904.1 | - | 2 | 504 | |||
BC038245.1 | - | 2 | 898 | |||
CB050274.1 | - | 2 | 179 |