Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286625.1 | NP_001273554.1 | 4 | 521 | ||
NM_001286626.1 | NP_001273555.1 | 3 | 528 | |||
NM_001286627.1 | NP_001273556.1 | 3 | 528 | |||
NM_006711.4 | NP_006702.1 | 4 | 582 | |||
NM_080594.3 | NP_542161.1 | 4 | 716 | |||
NR_104485.1 | 3 | 650 | ||||
XM_005255048.1 | XP_005255105.1 | 4 | 809 | |||
XM_005255049.3 | XP_005255106.1 | 4 | 2274 | |||
XM_017022874.1 | XP_016878363.1 | 4 | 1175 | |||
GenBank mRNA | AB445101.1 | - | 3 | 241 | ||
AF015608.1 | - | 3 | 255 | |||
AF274003.1 | - | 3 | 217 | |||
AK289955.1 | - | 5 | 476 | |||
AK293343.1 | - | 3 | 233 | |||
AK316132.1 | - | 3 | 518 | |||
AL520572.3 | - | 4 | 499 | |||
AL559988.3 | - | 3 | 303 | |||
BC001659.2 | - | 4 | 406 | 2959954 | ||
BC001838.2 | - | 4 | 406 | 2959954 | ||
BC108316.1 | - | 4 | 527 | 6151864 | ||
BG716139.1 | - | 4 | 424 | |||
BI819738.1 | - | 3 | 211 | |||
BQ059026.1 | - | 2 | 384 | |||
DA062271.1 | - | 4 | 514 | |||
DA728900.1 | - | 4 | 418 | |||
L37368.1 | - | 4 | 790 |