Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080480.2 | NP_001073949.1 | 2 | 351 | ||
XM_006714999.2 | XP_006715062.1 | 2 | 88 | |||
XM_006715000.3 | XP_006715063.1 | 2 | 289 | |||
XM_011514313.2 | XP_011512615.1 | 2 | 289 | |||
GenBank mRNA | AB305043.1 | - | 2 | 121 | ||
AK131269.1 | - | 2 | 314 | |||
AK304748.1 | - | 2 | 296 | |||
BC150650.1 | - | 2 | 297 | 9057021 | ||
BC150652.1 | - | 2 | 297 | 9057023 |