Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NKX6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152568.3 | NP_689781.1 | 2 | 294 | ||
XM_017013143.1 | XP_016868632.1 | 3 | 1006 | |||
GenBank mRNA | AK057898.1 | - | 2 | 294 |