Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010860.1 | NP_001010860.1 | 2 | 1746 | ||
GenBank mRNA | AK292571.1 | - | 2 | 2032 |