Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF567 SIRNA Assays ›
Gene Symbol:
ZNF567
Gene Name:
zinc finger protein 567
Gene Aliases:
-
Chromosome Location:
Chr.19: 36666961 - 36723548 on Build GRCh38
UniGene ID:
Hs.412517
Species:
Human
siRNA ID
HSS153021
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF567
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001300979.1
NP_001287908.1
6
522
NM_001322911.1
NP_001309840.1
5
395
NM_001322912.1
NP_001309841.1
5
447
NM_001322913.1
NP_001309842.1
6
526
NM_001322914.1
NP_001309843.1
6
698
NM_001322915.1
NP_001309844.1
7
594
NM_001322916.1
NP_001309845.1
6
470
NM_001322917.1
NP_001309846.1
6
474
NM_001322918.1
NP_001309847.1
7
598
NM_001322919.1
NP_001309848.1
6
694
NM_001322920.1
NP_001309849.1
7
646
NM_152603.4
NP_689816.2
5
399
XM_011526584.2
XP_011524886.1
4
326
XM_011526585.2
XP_011524887.1
4
326
XM_017026416.1
XP_016881905.1
5
658
XM_017026417.1
XP_016881906.1
8
1741
XM_017026418.1
XP_016881907.1
6
601
XM_017026419.1
XP_016881908.1
5
657
XM_017026420.1
XP_016881909.1
6
1510
XM_017026421.1
XP_016881910.1
5
621
GenBank mRNA
AK093034.1
-
5
331
AK126691.1
-
3
1421
AK298556.1
-
5
389
AK303326.1
-
6
506
AK313647.1
-
5
395
BC033849.2
-
4
373
4472579
BG253190.1
-
4
383
BX640706.1
-
6
511
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0