Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300979.1 | NP_001287908.1 | 6 | 863 | ||
NM_001322911.1 | NP_001309840.1 | 5 | 736 | |||
NM_001322912.1 | NP_001309841.1 | 5 | 788 | |||
NM_001322913.1 | NP_001309842.1 | 6 | 867 | |||
NM_001322914.1 | NP_001309843.1 | 6 | 1039 | |||
NM_001322915.1 | NP_001309844.1 | 7 | 935 | |||
NM_001322916.1 | NP_001309845.1 | 6 | 811 | |||
NM_001322917.1 | NP_001309846.1 | 6 | 815 | |||
NM_001322918.1 | NP_001309847.1 | 7 | 939 | |||
NM_001322919.1 | NP_001309848.1 | 6 | 1035 | |||
NM_001322920.1 | NP_001309849.1 | 7 | 987 | |||
NM_152603.4 | NP_689816.2 | 5 | 740 | |||
XM_011526584.2 | XP_011524886.1 | 4 | 667 | |||
XM_011526585.2 | XP_011524887.1 | 4 | 667 | |||
XM_017026416.1 | XP_016881905.1 | 5 | 999 | |||
XM_017026417.1 | XP_016881906.1 | 8 | 2082 | |||
XM_017026418.1 | XP_016881907.1 | 6 | 942 | |||
XM_017026419.1 | XP_016881908.1 | 5 | 998 | |||
XM_017026420.1 | XP_016881909.1 | 6 | 1851 | |||
XM_017026421.1 | XP_016881910.1 | 5 | 962 | |||
GenBank mRNA | AK093034.1 | - | 5 | 672 | ||
AK126691.1 | - | 3 | 1762 | |||
AK303326.1 | - | 6 | 847 | |||
AK313647.1 | - | 5 | 736 | |||
BC033849.2 | - | 4 | 714 | 4472579 | ||
BX640706.1 | - | 6 | 852 |