Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170743.3 | NP_734464.1 | 7 | 1058 | ||
NM_173064.2 | NP_775087.1 | 7 | 971 | |||
NM_173065.2 | NP_775088.1 | 6 | 927 | |||
XM_006710394.3 | XP_006710457.2 | 6 | 1122 | |||
XM_017000479.1 | XP_016855968.1 | 6 | 1087 | |||
GenBank mRNA | AF439325.1 | - | 7 | 1031 | ||
AJ534330.1 | - | 7 | 1044 | |||
AJ534331.1 | - | 6 | 913 | |||
AK160364.1 | - | 7 | 1034 | |||
AY129151.1 | - | 7 | 1031 | |||
AY129152.1 | - | 7 | 944 | |||
BC140872.1 | - | 7 | 1107 | 9021740 | ||
JN591376.1 | - | 7 | 1031 |