Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010862.2 | NP_001010862.2 | 2 | 517 | ||
NR_027139.1 | 2 | 517 | ||||
XM_006724577.3 | XP_006724640.1 | 2 | 578 | |||
XM_006724578.3 | XP_006724641.1 | 2 | 485 | |||
XM_017029315.1 | XP_016884804.1 | 2 | 920 | |||
GenBank mRNA | AK123122.1 | - | 2 | 581 | ||
AK304047.1 | - | 2 | 454 | |||
BC018073.1 | - | 2 | 461 | |||
BC032490.1 | - | 2 | 471 | |||
BC146899.1 | - | 2 | 238 | 9056827 | ||
BC146906.1 | - | 2 | 238 | 9056834 | ||
BX403408.2 | - | 2 | 465 | |||
DB151976.1 | - | 2 | 517 |