Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C10ORF128 SIRNA Assays ›
Gene Symbol:
C10orf128
Gene Name:
chromosome 10 open reading frame 128
Gene Aliases:
-
Chromosome Location:
Chr.10: 49154724 - 49188580 on Build GRCh38
UniGene ID:
Hs.385493
Species:
Human
siRNA ID
HSS153153
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C10orf128
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001010863.2
NP_001010863.1
3
255
NM_001288740.1
NP_001275669.1
3
255
NM_001288741.1
NP_001275670.1
3
255
NM_001288742.1
NP_001275671.1
3
255
NM_001288743.1
NP_001275672.1
3
255
XM_011539378.2
XP_011537680.1
3
435
XM_011539379.1
XP_011537681.1
3
435
XM_011539380.1
XP_011537682.1
3
435
XM_011539382.1
XP_011537684.1
3
435
XM_011539383.2
XP_011537685.1
3
435
XM_011539384.1
XP_011537686.1
3
435
XM_011539385.1
XP_011537687.1
3
435
XM_017015792.1
XP_016871281.1
3
435
XM_017015793.1
XP_016871282.1
3
435
XM_017015794.1
XP_016871283.1
3
435
XM_017015795.1
XP_016871284.1
3
435
XM_017015796.1
XP_016871285.1
3
435
XM_017015797.1
XP_016871286.1
3
435
GenBank mRNA
AK303891.1
-
3
253
AL524299.3
-
3
230
BC031641.1
-
3
362
BC037922.2
-
3
246
BC042857.1
-
3
246
BC047724.1
-
3
211
BC104438.1
-
3
217
BC104439.1
-
3
217
BU145920.1
-
3
167
DA366138.1
-
3
208
DC389158.1
-
3
256
DQ884399.1
-
3
191
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0