Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321522.1 | NP_001308451.1 | 5 | 534 | ||
NM_001321523.1 | NP_001308452.1 | 5 | 473 | |||
NM_001321524.1 | NP_001308453.1 | 5 | 481 | |||
NM_138374.2 | NP_612383.1 | 4 | 379 | |||
GenBank mRNA | AK295079.1 | - | 5 | 453 | ||
DC400704.1 | - | 4 | 351 |
Gene Symbol: ZNF525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003699.1 | 4 | 317 | |||
GenBank mRNA | DB221179.1 | - | 4 | 317 |