Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133498.2 | NP_598005.1 | 1 | 728 | ||
GenBank mRNA | AF353721.2 | - | 1 | 728 | ||
AY358387.1 | - | 1 | 710 | |||
BC047334.1 | - | 1 | 708 | 5163261 |